DNA-Dye NonTox Nucleic acid gel stain
Lieferant: APPLICHEM LIFESCIENCE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A9555.1000
A9555.1000EA 68.4 EUR InStock
A9555.1000
DNA-Dye NonTox Nucleic acid gel stain
Färbemittel Elektrophorese-Färbemittel
DNA-Dye NonTox is a highly sensitive, non toxic, fluorescent staining reagent for the detection of double-stranded DNA (dsDNA). The dye produces instant visualisation of DNA bands on gels upon blue light or UV illumination.
  • nonhazardous Ethidium bromide substitute
  • Loading buffer with highly sensitive DNA dye
  • Higher transfection rates since DNA structure and integrity is not affected
  • Ready to use
  • Contains three tracking dyes Bromophenol Blue, Xylene Cyanol FF, and Orange G for visually tracking the DNA migration during the electrophoresis process
DNA-Dye NonTox is ideal in terms of environmental safety by offering a non hazardous alternative to ethidium bromide. In addition, the dye included in DNA-Dye NonTox does not affect the structure and integrity of DNA.
Lieferumfang: Supplied in 6X DNA loading buffer to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge