Amino acid standards, physiological, acidics, neutrals, and basics, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A9906-1ML
A9906-1MLEA 89.8 EUR InStock
A9906-1ML
Amino acid standards, physiological, acidics, neutrals, and basics, analytical standard, Supelco®
Standards Lebensmittel-Standards
Amino acids and related compounds are 0,5 μmol/ml in 0,2 N lithium citrate, pH 2,2 containing 0,1% phenol and 2% thiodiglycol.
  • HPLC and gas chromatography (GC) suitable
The following components are included in the amino acid standard:
L-Alanine; Ammonium chloride; L-Arginine; L-Aspartic acid; L-Cystine; L-Glutamic acid; Glycine; L-Histidine; L-Isoleucine; L-Leucine; L-Lysine; L-Methionine; L-Phenylalanine; L-Proline; L-Serine; L-Threonine; L-Tyrosine; L-Valine; β-Alanine; L-α-Amino-n-butyric acid; γ-Amino-n-butyric acid; DL-β-Aminoisobutryic acid; L-Anserine; L-Carnosine; L-Citrulline; L-Creatinine; Cystathionine; Ethanolamine; L-Homocystine; δ-Hydroxylysine; Hydroxy-L-proline; 1-Methyl-L-histidine; 3-Methyl-L-histidine; L-Ornithine; Sarcosine; Taurine; L-Tryptophan; Urea
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge