Human β-Secretase (BACE1) Inhibitor Screening Kit (Fluorometric)
Lieferant: ABCAM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB283408-100
ABCAAB283408-100EA 930 EUR InStock
ABCAAB283408-100
Human β-Secretase (BACE1) Inhibitor Screening Kit (Fluorometric)
Tests Proteintests
Highly Sensitive Assay for screening potential BACE1 inhibitors.
  • Simple procedure takes one hour
  • Fast and convenient
  • The assay is sensitive, stable and high-throughput adaptable
β-Secretase (Beta-site amyloid precursor protein (APP) cleaving enzyme 1/BACE1), participates in the initial cleavage of amyloid precursor-APP generating two soluble N-terminal and C-terminal fragments. Another secretase isozyme, γ-Secretase, cleaves C-terminal fragment producing amyloid β.
Accumulation of amyloid β in brain results in the formation of amyloid plaques, which contribute in the development of Alzheimer’s disease (AD) in humans. Therefore, screening for novel and specific inhibitors of human β-Secretase (BACE1) has become critical in Alzheimer's disease-related drug discovery research.
Human β-Secretase (BACE1) inhibitor screening kit, human BACE1 cleaves a quenched substrate, specific for BACE1, generating a product with high fluorescence that can be measured at 435/500 nm. In the presence of BACE1 Inhibitor (control included in this kit), the enzymatic cleavage is impeded and could be used to screen for new inhibitor compounds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge