Angiotensin II (Ang II) (Human) ELISA Kit
Lieferant: ABCAM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB285306-96
ABCAAB285306-96EA 1430 EUR InStock
ABCAAB285306-96
Angiotensin II (Ang II) (Human) ELISA Kit
Tests ELISAs
Angiotensin II ELISA kit is a competitive ELISA assay for the quantitative measurement of Ang II in Human serum, plasma, tissue homogenates and other biological fluids.
  • Easy, convenient, and time-saving method to assay for human p53
  • The minimum detectable dose of p53 is typically less than 2 µg per sample
Angiotensin II acts directly on vascular smooth muscle as a potent vasoconstrictor, affects cardiac contractility and heart rate through its action on the sympathetic nervous system, and alters renal sodium and water absorption through its ability to stimulate the zona glomerulosa cells of the adrenal cortex to synthesize and secrete aldosterone. The microtiter plate provided in this kit has been pre-coated with the target antigen. During the reaction, the target in the sample or standard competes with a fixed amount of target on the solid phase supporter for sites on the Biotinylated Detection Antibody specific to target.
The unbound sample or excess conjugate are washed from the plate. HRP-Streptavidin (SABC) is added and incubated. Then TMB substrate solution is added to each well. The HRP enzymatic reaction is detected using TMB-substrate. Finally, an acidic stop solution terminates the enzymatic reaction. The color developed is measure at 450 nm. The concentration of target in the samples is then determined by comparing the OD of the samples to the standard curve.
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF,BLC, ENA-78, FGF-4, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF,GM-CSF, IFN-γ, Leptin (OB), MCP-1, MDC, MIP-1α, MIP-1 β, MIP-1δ, MMP-1, -2, -3, -10, PARC, RANTES, SCF, TARC, TGF-β, TIMP-1, TIMP-2, TNF-α, TNF-β, TPO, VEGF.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge