QuickDetect™ 1-3-β-D-glucan (Human) ELISA Kit
Lieferant: ABCAM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB287791-96
ABCAAB287791-96EA 954 EUR InStock
ABCAAB287791-96
QuickDetect™ 1-3-β-D-glucan (Human) ELISA Kit
Tests ELISAs
A rapid ELISA kit for quantify of 1-3-β-D-glucan in human serum, plasma, urine, cell culture samples.
  • Easy, convenient and time-saving method to measure the level of 1-3-β-D-glucan in human Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 0,8 to 50 pg/ml
  • Sensitivity: <0,1 pg/ml
β-Glucans (beta-glucans) comprise a group of β-D-glucose polysaccharides naturally occurring in the cell walls of cereals, bacteria, and fungi, with significantly differing physicochemical properties dependent on source. Typically, β-glucans form a linear backbone with 1-3 β-glycosidic bonds but vary with respect to molecular mass, solubility, viscosity, branching structure, and gelation properties, causing diverse physiological effects in animals. BioVision’s 1-3-β-DG ELISA kit is a sandwich ELISA assay for the quantitative measurement of Human 1-3-β-DG in serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Human 1-3-β-DG captured from the samples.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge