Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

QuickDetect™ Acetylcholine (ACh) (Mouse) ELISA Kit

Lieferant: ABCAM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


AB287812-96
ABCAAB287812-96EA 954 EUR InStock
ABCAAB287812-96
QuickDetect™ Acetylcholine (ACh) (Mouse) ELISA Kit
Tests ELISAs
A rapid ELISA kit to quantify Acetylcholine in mouse serum, plasma, urine, cell culture samples.

  • Easy, convenient and time-saving method to measure the level of Acetylcholine in mouse Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 2,4 to 140 pg/ml
  • Sensitivity: <0,5 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed

Acetylcholine (ACh) is an organic chemical that functions in the brain and body of many types of animals, including Mouses, as a neurotransmitter-a chemical released by nerve cells to send signals to other cells. Acetylcholine is also used as a neurotransmitter in the autonomic nervous system, both as an internal transmitter for the sympathetic nervous system and as the final product released by the parasympathetic nervous system. In the brain, acetylcholine functions as a neurotransmitter and as a neuromodulator. The brain contains a number of cholinergic areas, each with distinct functions. They play an important role in arousal, attention, memory and motivation. BioVision’s Acetylcholine ELISA kit is a sandwich ELISA assay for the quantitative measurement of ACh in mouse serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Acetylcholine captured from the samples.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge