Protein G Spin Antibody Purification Kit
Lieferant: ABCAM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB288095-10
ABCAAB288095-10EA 340 EUR InStock
ABCAAB288095-10
Protein G Spin Antibody Purification Kit
Tests Proteintests
Kit helps purify antibodies from serum, ascites and cell culture medium from various species.
  • Ready-to-use pre-packed spin columns of 0,1 ml bead volume in 20% ethanol
  • High binging capacity: Binding of IgG ≥30 mg human, mouse or rabbit IgG mg/ml protein G agarose
Protein G beads are widely used for IgG purification for their ability to bind selectively immunoglobulins. The protein G is a genetically engineered protein containing three Ig-binding regions of native protein G. The cell wall binding region, albumin binding region and other non-specific regions have been eliminated from the recombinant protein G to ensure the maximum specific IgG binding. The Hi-Bind™ protein G-Agarose beads display high chemical and physical stability as well as high flow rate, hydrophilicity and high gel strength. The protein G spin antibody purification kit is a simple, ready to use kit containing all necessary buffers and pre-packed columns to purify up to 3 mg of IgG per sample in ten samples (30 mg IgG total). The kit can be used to purify antibodies in serum, ascites and cell culture medium from various species such as human, mouse, rat, goat and rabbit.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge