A19396.14
A19396.22
AB125716.0100
A19396.14EA
19.1
EUR
InStock
A19396.14
A19396.22
ABCRAB125716.0100
5-Brom-o-anisaldehyd ≥98%
5-Brom-o-anisaldehyd
Purity: 98+%
CAS No.: 25016-01-7
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 114-118°
Beilstein Reference: 508840