A19206.0E
A19206.36
AB125754.0500
A19206.0EEA
413
EUR
InStock
A19206.0E
A19206.36
ABCRAB125754.0500
2-tert-Butylhydrochinon ≥97%
2-tert-Butylhydrochinon
Purity: 97%
CAS No.: 1948-33-0
Molecular Formula: C10H14O2
Formula Weight: 166.22
Flash Point: 171°(339°F)
Melting Point: 126-129°
Boiling Point: 295°
Beilstein Reference: 637923
UN No.: 3077
MDL No.: MFCD00002344