L14035.22
L14035.14
AB173857.0100
L14035.22EA
204
EUR
InStock
L14035.22
L14035.14
ABCRAB173857.0100
Dimethoxymethylvinylsilan ≥97%
Dimethoxymethylvinylsilan
Purity: 97%
CAS No.: 16753-62-1
Molecular Formula: C5H12O2Si
Formula Weight: 132.23
Flash Point: 5°(41°F)
Refractive Index: 1.3950
Boiling Point: 102-104°
Beilstein Reference: 3043765
UN No.: 1993
MDL No.: MFCD00129170