L20134.06
L20134.14
AB174305.0025
L20134.06EA
154
EUR
InStock
L20134.06
L20134.14
ABCRAB174305.0025
1-Brom-2-butin ≥98%
1-Brom-2-butin
Purity: 98%
CAS No.: 3355-28-0
Molecular Formula: CH3C≡CCH2Br
Formula Weight: 132.99
Flash Point: 36°(96°F)
Refractive Index: 1.5080
Boiling Point: 123-125°
Beilstein Reference: 605306
UN No.: 1993
MDL No.: MFCD00190233