Biotin labelling kit
Lieferant: ABNOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KA0003 KA0004
ABNOKA0003EA 303 EUR InStock
ABNOKA0003 ABNOKA0004
Biotin labelling kit
Tests Proteintests
Biotin Labelling Kit is used for simple and rapid preparation of biotin-labeled antibodies and proteins.
Achtung! Bitte beachten: Store at 4 °C. For research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge