Estradiol (Human) ELISA kit
Lieferant: ABNOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KA6702.1KIT
ABNOKA6702.1KITEA 359 EUR InStock
ABNOKA6702.1KIT
Estradiol (Human) ELISA kit
Tests ELISAs
Estradiol (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of estradiol in serum and plasma.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge