Cytomegalovirus IgM ELISA kit
Lieferant: ABNOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KA6725.1KIT
ABNOKA6725.1KITEA 359 EUR InStock
ABNOKA6725.1KIT
Cytomegalovirus IgM ELISA kit
Tests ELISAs
Cytomegalovirus IgM ELISA Kit is a qualitative determination of IgM class antibodies against Cytomegalovirus (CMV) in human serum or plasma.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge