The IgM (Monkey) ELISA kit
Lieferant: ABNOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KA6751.1KIT
ABNOKA6751.1KITEA 1000 EUR InStock
ABNOKA6751.1KIT
The IgM (Monkey) ELISA kit
Tests ELISAs
The IgM (Monkey) ELISA Kit is used for determination of IgM in old world monkey serum or plasma.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge