Methoxy-PEG ELISA kit
Lieferant: ABNOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KA6893.1KIT
ABNOKA6893.1KITEA 1140 EUR InStock
ABNOKA6893.1KIT
Methoxy-PEG ELISA kit
Tests ELISAs
Methoxy-PEG ELISA Kit is a sandwich-type ELISA and use two different monoclonal antibodies. The capture antibody recognizes the polyoxyethylene backbone. The other is specific for the terminal methoxy group of mPEGs and mPEGylated proteins.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge