Biotinylated Human 4-1BB Ligand, Fc,Avitag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 ( Accession P41273-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human 4-1BB Ligand, Fc,Avitag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 ( Accession P41273-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human 4-1BB Ligand, Fc,Avitag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 ( Accession P41273-1). 1 * 200 µG ACRB41L-H82F9200UG 1 * 200 µG
Hersteller-Artikelnummer:: 41L-H82F9200UG
Lokale Artikelnummer:: ACRB41L-H82F9200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder