Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 1 mg ACRBACA-H424X-1MG 1 * 1 mg
Hersteller-Artikelnummer:: ACA-H424X-1MG
Lokale Artikelnummer:: ACRBACA-H424X-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder