Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession P05230-1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession P05230-1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession P05230-1). 1 * 200 µG ACRBAFF-H4116200UG 1 * 200 µG
Hersteller-Artikelnummer:: AFF-H4116200UG
Lokale Artikelnummer:: ACRBAFF-H4116200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder