Recombinat Protein,Human Angiopoietin-2 ANGPT2 (275-496) Protein, His Tag,Human Angiopoietin-2 (275-496), His Tag (AN2-H5242) is ex pressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This protein carries a 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-2 ANGPT2 (275-496) Protein, His Tag,Human Angiopoietin-2 (275-496), His Tag (AN2-H5242) is ex pressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This protein carries a 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Angiopoietin-2 ANGPT2 (275-496) Protein, His Tag,Human Angiopoietin-2 (275-496), His Tag (AN2-H5242) is ex pressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This protein carries a 1 * 100 µG ACRBAN2-H5242100UG 1 * 100 µG
Hersteller-Artikelnummer:: AN2-H5242100UG
Lokale Artikelnummer:: ACRBAN2-H5242100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder