Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg ACRBAN3-H5226-1MG 1 * 1 mg
Hersteller-Artikelnummer:: AN3-H5226-1MG
Lokale Artikelnummer:: ACRBAN3-H5226-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder