Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg ACRBAP1-H5225-1MG 1 * 1 mg
Hersteller-Artikelnummer:: AP1-H5225-1MG
Lokale Artikelnummer:: ACRBAP1-H5225-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder