Human B7-H3, Fc Tag (B73-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession Q5ZPR3-2). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human B7-H3, Fc Tag (B73-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession Q5ZPR3-2). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human B7-H3, Fc Tag (B73-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession Q5ZPR3-2). 1 * 1 mg ACRBB73-H5253-1MG 1 * 1 mg
Hersteller-Artikelnummer:: B73-H5253-1MG
Lokale Artikelnummer:: ACRBB73-H5253-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder