Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568). 1 * 1 mg ACRBB75-H52H0-1MG 1 * 1 mg
Hersteller-Artikelnummer:: B75-H52H0-1MG
Lokale Artikelnummer:: ACRBB75-H52H0-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder