Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 200 µG ACRBBAF-H82Q2200UG 1 * 200 µG
Hersteller-Artikelnummer:: BAF-H82Q2200UG
Lokale Artikelnummer:: ACRBBAF-H82Q2200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder