Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 µG ACRBC5A-M51H4100UG 1 * 100 µG
Hersteller-Artikelnummer:: C5A-M51H4100UG
Lokale Artikelnummer:: ACRBC5A-M51H4100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder