Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H52 26) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein car 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H52 26) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein car 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Carbonic Anhydrase IX CA9 (38-414) Protein, His Tag,Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H52 26) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession NP001207).This protein car 1 * 1 mg ACRBCA9-H5226-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CA9-H5226-1MG
Lokale Artikelnummer:: ACRBCA9-H5226-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder