Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession NP001234.2). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession NP001234.2). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession NP001234.2). 1 * 100 µG ACRBCD0-H5250100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD0-H5250100UG
Lokale Artikelnummer:: ACRBCD0-H5250100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder