Recombinat Protein,Human CD31 PECAM-1 Protein, His Tag,Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession P16284-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD31 PECAM-1 Protein, His Tag,Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession P16284-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD31 PECAM-1 Protein, His Tag,Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession P16284-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG ACRBCD1-H5224100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD1-H5224100UG
Lokale Artikelnummer:: ACRBCD1-H5224100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder