Human CD52, Fc Tag (CD2-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession P31358-1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human CD52, Fc Tag (CD2-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession P31358-1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human CD52, Fc Tag (CD2-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession P31358-1). 1 * 1 mg ACRBCD2-H525A-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD2-H525A-1MG
Lokale Artikelnummer:: ACRBCD2-H525A-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder