Biotinylated Human CD52, His,Avitag (CD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accessio n P31358-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD52, His,Avitag (CD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accessio n P31358-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD52, His,Avitag (CD2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accessio n P31358-1). 1 * 25 µG ACRBCD2-H82E8-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CD2-H82E8-25UG
Lokale Artikelnummer:: ACRBCD2-H82E8-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder