Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 1 mg ACRBCD3-H5255-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD3-H5255-1MG
Lokale Artikelnummer:: ACRBCD3-H5255-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder