Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 20 G
Lieferant: ACROBIOSYSTEMS
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 20 G
PRODUKT
Erweitert / Kompakt
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 20 µG ACRBCD3-H52H4-20UG 1 * 20 µG
Hersteller-Artikelnummer:: CD3-H52H4-20UG
Lokale Artikelnummer:: ACRBCD3-H52H4-20UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder