Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 1 mg ACRBCD4-H524C-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD4-H524C-1MG
Lokale Artikelnummer:: ACRBCD4-H524C-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder