Human CD24, Fc Tag (CD4-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human CD24, Fc Tag (CD4-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human CD24, Fc Tag (CD4-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 µG ACRBCD4-H5254100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD4-H5254100UG
Lokale Artikelnummer:: ACRBCD4-H5254100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder