Recombinat Protein,Biotinylated Human CD84 SLAMF5 Protein, His,Avitag,Biotinylated Human CD84, His,Avitag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession Q9UIB8-3).This protein carries a polyhisti e 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human CD84 SLAMF5 Protein, His,Avitag,Biotinylated Human CD84, His,Avitag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession Q9UIB8-3).This protein carries a polyhisti e 1 * 25 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Biotinylated Human CD84 SLAMF5 Protein, His,Avitag™,Biotinylated Human CD84, His,Avitag (CD4-H82E5) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession Q9UIB8-3).This protein carries a polyhisti e 1 * 25 µG ACRBCD4-H82E5-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CD4-H82E5-25UG
Lokale Artikelnummer:: ACRBCD4-H82E5-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder