Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accessio n P25063-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accessio n P25063-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD24, His,Avitag (CD4-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accessio n P25063-1). 1 * 25 µG ACRBCD4-H82E9-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CD4-H82E9-25UG
Lokale Artikelnummer:: ACRBCD4-H82E9-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder