Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg ACRBCD5-H5225-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD5-H5225-1MG
Lokale Artikelnummer:: ACRBCD5-H5225-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder