Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 3 43 (Accession NP006496.4). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 3 43 (Accession NP006496.4). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human CD155, Mouse IgG2a Fc Tag, low endotoxin (CD5-H5254) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 3 43 (Accession NP006496.4). 1 * 100 µG ACRBCD5-H5254100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD5-H5254100UG
Lokale Artikelnummer:: ACRBCD5-H5254100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder