Recombinat Protein,Human CD5 Protein, His Tag,Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession NP055022).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD5 Protein, His Tag,Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession NP055022).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD5 Protein, His Tag,Human CD5, His Tag (CD5-H52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession NP055022).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg ACRBCD5-H52H5-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD5-H52H5-1MG
Lokale Artikelnummer:: ACRBCD5-H52H5-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder