Biotinylated Human CD155, His,Avitag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Access ion NP006496.4). 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD155, His,Avitag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Access ion NP006496.4). 1 * 200 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD155, His,Avitag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Access ion NP006496.4). 1 * 200 µG ACRBCD5-H82E3200UG 1 * 200 µG
Hersteller-Artikelnummer:: CD5-H82E3200UG
Lokale Artikelnummer:: ACRBCD5-H82E3200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder