Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession Q0 MSE6-1). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession Q0 MSE6-1). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession Q0 MSE6-1). 1 * 100 µG ACRBCD5-R52H5100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD5-R52H5100UG
Lokale Artikelnummer:: ACRBCD5-R52H5100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder