Cynomolgus Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Acc ession G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus. 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Acc ession G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus. 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Beschreibung Katalognummer Verpackungseinheit Menge
Cynomolgus Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Acc ession G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus. 1 * 1 mg ACRBCD6-C5254-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD6-C5254-1MG
Lokale Artikelnummer:: ACRBCD6-C5254-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder