Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 100 G
PRODUKT
Erweitert / Kompakt
Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 100 µG ACRBCD6-C52H5100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD6-C52H5100UG
Lokale Artikelnummer:: ACRBCD6-C52H5100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder