Recombinat Protein,Human ALCAM Protein, His Tag,Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It cont ains AA Trp 28 - Ala 526 (Accession Q13740-1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 200 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human ALCAM Protein, His Tag,Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It cont ains AA Trp 28 - Ala 526 (Accession Q13740-1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 200 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human ALCAM Protein, His Tag,Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It cont ains AA Trp 28 - Ala 526 (Accession Q13740-1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 200 µG ACRBCD6-H5228200UG 1 * 200 µG
Hersteller-Artikelnummer:: CD6-H5228200UG
Lokale Artikelnummer:: ACRBCD6-H5228200UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder