Recombinat Protein,Human EMMPRIN CD147 Protein, Fc Tag,Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293) . It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human EMMPRIN CD147 Protein, Fc Tag,Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293) . It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human EMMPRIN CD147 Protein, Fc Tag,Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293) . It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a human IgG1 Fc tag at the C-terminus. s 1 * 100 µG ACRBCD7-H5259100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD7-H5259100UG
Lokale Artikelnummer:: ACRBCD7-H5259100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder