Biotinylated Human CD47, His,Avitag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on NP942088). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human CD47, His,Avitag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on NP942088). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human CD47, His,Avitag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accessi on NP942088). 1 * 25 µG ACRBCD7-H82E9-25UG 1 * 25 µG
Hersteller-Artikelnummer:: CD7-H82E9-25UG
Lokale Artikelnummer:: ACRBCD7-H82E9-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder