Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630 (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630 (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630  (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg ACRBCD8-H5247-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD8-H5247-1MG
Lokale Artikelnummer:: ACRBCD8-H5247-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder