Human CD38, Fc Tag (CD8-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession P28907-1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human CD38, Fc Tag (CD8-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession P28907-1). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human CD38, Fc Tag (CD8-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession P28907-1). 1 * 1 mg ACRBCD8-H5255-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CD8-H5255-1MG
Lokale Artikelnummer:: ACRBCD8-H5255-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder