Human Cynomolgus Rhesus macaque CD28, Mouse IgG2a Fc Tag, low endotoxin (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolg 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human Cynomolgus Rhesus macaque CD28, Mouse IgG2a Fc Tag, low endotoxin (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolg 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human Cynomolgus Rhesus macaque CD28, Mouse IgG2a Fc Tag, low endotoxin (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolg 1 * 100 µG ACRBCD8-H52A5100UG 1 * 100 µG
Hersteller-Artikelnummer:: CD8-H52A5100UG
Lokale Artikelnummer:: ACRBCD8-H52A5100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder